siRNA Details
| Gene Symbol: Pomt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153415.3 | NP_700464.2 | 3 | 854 | ||
| GenBank mRNA | AF246235.1 | - | 3 | 458 | ||
| AK043573.1 | - | 3 | 555 | |||
| AK043713.1 | - | 3 | 396 | |||
| AK050915.1 | - | 3 | 423 | |||
| AK076813.1 | - | 3 | 856 | |||
| AK085703.1 | - | 3 | 922 | |||
| AY090481.1 | - | 3 | 854 | |||
| AY090482.1 | - | 3 | 454 | |||
| BC052045.1 | - | 3 | 426 | 6402328 | ||
| BC062965.1 | - | 3 | 426 | |||
| BC079589.1 | - | 3 | 424 | |||

Pomt2
Genomic Map