siRNA Details
| Gene Symbol: Pomt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153415.3 | NP_700464.2 | 10 | 1616 | ||
| GenBank mRNA | AF246235.1 | - | 10 | 1220 | ||
| AK043573.1 | - | 10 | 1317 | |||
| AK043713.1 | - | 10 | 1158 | |||
| AK050915.1 | - | 10 | 1185 | |||
| AK076813.1 | - | 10 | 1618 | |||
| AY090481.1 | - | 10 | 1616 | |||
| AY090482.1 | - | 10 | 1216 | |||
| BC052045.1 | - | 10 | 1188 | 6402328 | ||
| BC062965.1 | - | 10 | 1188 | |||
| BC079589.1 | - | 10 | 1186 | |||

Pomt2
Genomic Map