siRNA Details
| Gene Symbol: Ammecr1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242430.1 | NP_001229359.1 | 8 | 2913 | ||
| NM_153515.4 | NP_705735.1 | 8 | 2712 | |||
| GenBank mRNA | BC012310.1 | - | 8 | 2445 | ||
| BC034661.1 | - | 8 | 2453 | 5137474 | ||
| BC056948.1 | - | 8 | 2656 | 6827318 | ||

Ammecr1l
Genomic Map