siRNA Details
| Gene Symbol: Lrrc41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK052762.1 | - | 4 | 904 | ||
| AK154613.1 | - | 4 | 1050 | |||
| AK163171.1 | - | 4 | 997 | |||
| AK170641.1 | - | 4 | 1036 | |||
| AK202388.1 | - | 1 | 66 | |||
| BC005765.1 | - | 3 | 772 | |||
| BC022976.1 | - | 4 | 982 | 5362925 | ||
| BC024940.1 | - | 4 | 980 | |||
| BC071258.1 | - | 4 | 915 | |||

Lrrc41
Genomic Map