siRNA Details
| Gene Symbol: Fam149a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153535.2 | NP_705763.1 | 7 | 1958 | ||
| GenBank mRNA | AK034229.1 | - | 7 | 1185 | ||
| AK038552.1 | - | 7 | 1173 | |||
| AK140440.1 | - | 7 | 1958 | |||
| AK141607.1 | - | 7 | 1901 | |||
| AK220518.1 | - | 7 | 1194 | |||
| BC035537.1 | - | 7 | 1849 | 5369152 | ||

Fam149a
Genomic Map