siRNA Details
Gene Symbol: Evi5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039578.3 | NP_001034667.3 | 7 | 1045 | ||
GenBank mRNA | AK045240.1 | - | 7 | 1046 | ||
BC042653.1 | - | 7 | 1025 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Evi5l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039578.3 | NP_001034667.3 | 7 | 1045 | ||
GenBank mRNA | AK045240.1 | - | 7 | 1046 | ||
BC042653.1 | - | 7 | 1025 |