siRNA Details
| Gene Symbol: Evi5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039578.3 | NP_001034667.3 | 7 | 1120 | ||
| GenBank mRNA | AK045240.1 | - | 7 | 1121 | ||
| AK180516.1 | - | 1 | 75 | |||
| BC042653.1 | - | 7 | 1100 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Evi5l| Gene Symbol: Evi5l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039578.3 | NP_001034667.3 | 7 | 1120 | ||
| GenBank mRNA | AK045240.1 | - | 7 | 1121 | ||
| AK180516.1 | - | 1 | 75 | |||
| BC042653.1 | - | 7 | 1100 | |||