siRNA Details
| Gene Symbol: Brinp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145807.1 | NP_001139279.1 | 3 | 461 | ||
| NM_153539.3 | NP_705767.3 | 3 | 819 | |||
| GenBank mRNA | AK046878.1 | - | 3 | 820 | ||
| AK083233.1 | - | 4 | 510 | |||
| BC030498.1 | - | 2 | 437 | 5400263 | ||
| BC099956.1 | - | 3 | 796 | 30101722 | ||
| CX199744.1 | - | 3 | 461 | |||

Brinp3
Genomic Map