siRNA Details
| Gene Symbol: Aldh1l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153543.2 | NP_705771.2 | 3 | 386 | ||
| GenBank mRNA | AK052361.1 | - | 3 | 277 | ||
| AK086413.1 | - | 3 | 328 | |||
| AK141954.1 | - | 3 | 348 | |||
| BC034531.1 | - | 3 | 308 | 4188055 | ||
| CJ130150.1 | - | 3 | 386 | |||

Aldh1l2
Genomic Map