siRNA Details
| Gene Symbol: Lrrc45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153545.2 | NP_705773.2 | 18 | 2350 | ||
| GenBank mRNA | AK081566.1 | - | 17 | 2278 | ||
| AK153361.1 | - | 18 | 2351 | |||
| BC023296.1 | - | 18 | 2330 | 3662961 | ||
| CN539620.1 | - | 3 | 600 | |||

Lrrc45
Genomic Map