siRNA Details
| Gene Symbol: E430025E21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153548.2 | NP_705776.2 | 5 | 664 | ||
| GenBank mRNA | AK128970.1 | - | 5 | 679 | ||
| AK215294.1 | - | 4 | 304 | |||
| BC067035.1 | - | 5 | 664 | 30532658 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

E430025E21Rik| Gene Symbol: E430025E21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153548.2 | NP_705776.2 | 5 | 664 | ||
| GenBank mRNA | AK128970.1 | - | 5 | 679 | ||
| AK215294.1 | - | 4 | 304 | |||
| BC067035.1 | - | 5 | 664 | 30532658 | ||