siRNA Details
Gene Symbol: BC029214 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153557.1 | NP_705785.1 | 3 | 381 | ||
GenBank mRNA | AK046066.1 | - | 2 | 311 | ||
BC029214.1 | - | 3 | 381 | 4161391 |

More Information
Other Species Targeted

Gene Ontology Categories:
