siRNA Details
| Gene Symbol: Gbp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153564.2 | NP_705792.2 | 4, 5 | 494 | ||
| GenBank mRNA | AF422243.1 | - | 4, 5 | 476 | ||
| AK030414.1 | - | 4, 5 | 494 | |||
| AK087083.1 | - | 582 | ||||
| BC058555.1 | - | 4, 5 | 528 | 5293575 | ||

Gbp5
Genomic Map