siRNA Details
| Gene Symbol: Lrrc66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153568.1 | NP_705796.1 | 2 | 276 | ||
| GenBank mRNA | AK051875.1 | - | 2 | 283 | ||
| BC031901.1 | - | 2 | 276 | 5362830 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrrc66| Gene Symbol: Lrrc66 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153568.1 | NP_705796.1 | 2 | 276 | ||
| GenBank mRNA | AK051875.1 | - | 2 | 283 | ||
| BC031901.1 | - | 2 | 276 | 5362830 | ||