siRNA Details
Gene Symbol: Lrrc66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153568.1 | NP_705796.1 | 5 | 2357 | ||
GenBank mRNA | AK051875.1 | - | 5 | 2364 | ||
BC031901.1 | - | 5 | 2357 | 5362830 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Lrrc66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153568.1 | NP_705796.1 | 5 | 2357 | ||
GenBank mRNA | AK051875.1 | - | 5 | 2364 | ||
BC031901.1 | - | 5 | 2357 | 5362830 |