siRNA Details
| Gene Symbol: Paox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153783.4 | NP_722478.2 | 7 | 1505 | ||
| GenBank mRNA | AF226656.2 | - | 7 | 1524 | ||
| AK010469.1 | - | 6 | 1155 | |||
| AK030664.1 | - | 7 | 1506 | |||
| AK159137.1 | - | 5 | 792 | |||
| AM049402.1 | - | 7 | 1497 | |||
| BC033913.1 | - | 6 | 769 | |||
| BC082783.1 | - | 7 | 1504 | 6475809 | ||

Paox
Genomic Map