siRNA Details
| Gene Symbol: Acap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153788.3 | NP_722483.2 | 2, 3 | 320 | ||
| GenBank mRNA | AK133473.1 | - | 2, 3 | 323 | ||
| AK138736.1 | - | 2, 3 | 312 | |||
| AK154449.1 | - | 2, 3 | 323 | |||
| AK155001.1 | - | 2, 3 | 341 | |||
| BC031462.1 | - | 2, 3 | 320 | 4017986 | ||

Acap1
Genomic Map