siRNA Details
| Gene Symbol: Scarf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153790.2 | NP_722485.1 | 7 | 1340 | ||
| GenBank mRNA | AB024432.1 | - | 7 | 1307 | ||
| AF522197.1 | - | 7 | 1341 | |||
| AK142378.1 | - | 7 | 1308 | |||
| AK143208.1 | - | 7 | 1183 | |||
| AK170331.1 | - | 3 | 394 | |||
| BC059226.1 | - | 7 | 1272 | |||
| BC060084.1 | - | 7 | 1272 | |||
| BC139758.1 | - | 7 | 1293 | 40047931 | ||

Scarf2
Genomic Map