siRNA Details
| Gene Symbol: Peo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153796.3 | NP_722491.2 | 1 | 1105 | ||
| GenBank mRNA | AK146244.1 | - | 1 | 999 | ||
| AY059385.1 | - | 1 | 567 | |||
| BC071195.1 | - | 1 | 1105 | 5100486 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Peo1| Gene Symbol: Peo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153796.3 | NP_722491.2 | 1 | 1105 | ||
| GenBank mRNA | AK146244.1 | - | 1 | 999 | ||
| AY059385.1 | - | 1 | 567 | |||
| BC071195.1 | - | 1 | 1105 | 5100486 | ||