siRNA Details
| Gene Symbol: Disp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_170593.3 | NP_733481.1 | 8 | 1190 | ||
| GenBank mRNA | AK035975.1 | - | 5 | 1879 | ||
| AK038938.1 | - | 7 | 2638 | |||
| AY150699.1 | - | 8 | 991 | |||
| BC118526.1 | - | 8 | 985 | 40099214 | ||

Disp2
Genomic Map