siRNA Details
| Gene Symbol: Fam134a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_170755.2 | NP_739561.2 | 1, 2 | 340 | ||
| GenBank mRNA | AK034667.1 | - | 1, 2 | 341 | ||
| AK160400.1 | - | 1, 2 | 294 | |||
| AK165957.1 | - | 1, 2 | 319 | |||
| AK172093.1 | - | 1, 2 | 337 | |||
| BC070406.1 | - | 1, 2 | 332 | 5707319 | ||

Fam134a
Genomic Map