siRNA Details
| Gene Symbol: Fam134a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_170755.2 | NP_739561.2 | 9 | 2318 | ||
| GenBank mRNA | AK034667.1 | - | 9 | 2321 | ||
| AK152268.1 | - | 9 | 2033 | |||
| AK158940.1 | - | 9 | 2087 | |||
| AK160400.1 | - | 9 | 2272 | |||
| AK165957.1 | - | 10 | 2185 | |||
| AK172093.1 | - | 9 | 2315 | |||
| AK218799.1 | - | 1 | 170 | |||
| BC026519.1 | - | 6 | 1558 | |||
| BC038286.1 | - | 6 | 1593 | 4221441 | ||
| BC070406.1 | - | 10 | 2199 | 5707319 | ||

Fam134a
Genomic Map