siRNA Details
| Gene Symbol: Ccdc186 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_170757.1 | NP_739563.1 | 1 | 186 | ||
| GenBank mRNA | AF349751.1 | - | 1 | 186 | ||
| AK041398.1 | - | 2 | 373 | |||
| AK054204.1 | - | 2 | 387 | |||
| BC064039.1 | - | 2 | 349 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc186| Gene Symbol: Ccdc186 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_170757.1 | NP_739563.1 | 1 | 186 | ||
| GenBank mRNA | AF349751.1 | - | 1 | 186 | ||
| AK041398.1 | - | 2 | 373 | |||
| AK054204.1 | - | 2 | 387 | |||
| BC064039.1 | - | 2 | 349 | |||