siRNA Details
| Gene Symbol: Pddc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK088159.1 | - | 7, 8 | 697 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pddc1| Gene Symbol: Pddc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK088159.1 | - | 7, 8 | 697 | ||