siRNA Details
| Gene Symbol: Zc3h3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172121.1 | NP_742119.1 | 12 | 2847 | ||
| GenBank mRNA | AJ516034.1 | - | 12 | 2847 | ||
| AK138281.1 | - | 12 | 2855 | |||
| BC038813.1 | - | 11 | 1784 | |||
| BC049953.1 | - | 11 | 1703 | |||
| BC060682.1 | - | 12 | 2846 | 6416246 | ||
| BC111448.1 | - | 4 | 169 | |||

Zc3h3
Genomic Map