siRNA Details
| Gene Symbol: Pdxk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172134.2 | NP_742146.1 | 3 | 325 | ||
| GenBank mRNA | AK039194.1 | - | 3 | 307 | ||
| AK080846.1 | - | 3 | 301 | |||
| AK145470.1 | - | 3 | 289 | |||
| AK166078.1 | - | 3 | 308 | |||
| AK166464.1 | - | 3 | 290 | |||
| BC027745.1 | - | 3 | 362 | 5064695 | ||
| BC032171.1 | - | 3 | 296 | |||
| BI143426.1 | - | 3 | 354 | |||

Pdxk
Genomic Map