siRNA Details
| Gene Symbol: Pdxk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172134.2 | NP_742146.1 | 11 | 2258 | ||
| GenBank mRNA | AK039194.1 | - | 11 | 2238 | ||
| AK191960.1 | - | 1 | 300 | |||
| AK206366.1 | - | 1 | 126 | |||
| BC027745.1 | - | 11 | 2297 | 5064695 | ||
| BC032171.1 | - | 11 | 2229 | |||

Pdxk
Genomic Map