siRNA Details
| Gene Symbol: DSCR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005867.3 | NP_005858.1 | 3 | 986 | ||
| GenBank mRNA | AB000099.1 | - | 3 | 986 | ||
| BC069729.1 | - | 3 | 948 | 7262631 | ||
| DQ179113.1 | - | 3 | 936 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DSCR4| Gene Symbol: DSCR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005867.3 | NP_005858.1 | 3 | 986 | ||
| GenBank mRNA | AB000099.1 | - | 3 | 986 | ||
| BC069729.1 | - | 3 | 948 | 7262631 | ||
| DQ179113.1 | - | 3 | 936 | |||