siRNA Details
| Gene Symbol: Mfsd9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172499.2 | NP_766087.1 | 6 | 1857 | ||
| GenBank mRNA | AK029874.1 | - | 6 | 1858 | ||
| AK169911.1 | - | 7 | 2017 | |||
| BC057633.1 | - | 6 | 1863 | 4035998 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mfsd9| Gene Symbol: Mfsd9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172499.2 | NP_766087.1 | 6 | 1857 | ||
| GenBank mRNA | AK029874.1 | - | 6 | 1858 | ||
| AK169911.1 | - | 7 | 2017 | |||
| BC057633.1 | - | 6 | 1863 | 4035998 | ||