siRNA Details
| Gene Symbol: Syne3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042699.2 | NP_001036164.1 | 8 | 1479 | ||
| NM_172500.3 | NP_766088.2 | 6 | 1124 | |||
| GenBank mRNA | AK029216.1 | - | 6 | 1126 | ||
| BC099694.1 | - | 8 | 1479 | 4507746 | ||

Syne3
Genomic Map