siRNA Details
| Gene Symbol: Syne3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042699.2 | NP_001036164.1 | 19 | 4627 | ||
| NM_172500.3 | NP_766088.2 | 17 | 4272 | |||
| GenBank mRNA | AK029216.1 | - | 17 | 4276 | ||
| AK030542.1 | - | 6 | 2271 | |||
| AK166152.1 | - | 6 | 2139 | |||
| BC063771.1 | - | 1 | 890 | |||

Syne3
Genomic Map