siRNA Details
| Gene Symbol: Kctd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172509.3 | NP_766097.1 | 4 | 1010 | ||
| GenBank mRNA | AK029942.1 | - | 4 | 986 | ||
| AK034583.1 | - | 4 | 986 | |||
| AK083583.1 | - | 4 | 1011 | |||
| AK140606.1 | - | 4 | 985 | |||
| AK162568.1 | - | 4 | 1039 | |||
| AK178565.1 | - | 1 | 129 | |||
| AK195855.1 | - | 1 | 129 | |||
| BC051544.1 | - | 3 | 553 | |||
| BC139070.1 | - | 4 | 912 | 8862092 | ||
| BC141201.1 | - | 4 | 912 | 9055813 | ||

Kctd7
Genomic Map