siRNA Details
| Gene Symbol: Gabpb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029885.1 | NP_084161.1 | 10 | 1717 | ||
| NM_172512.2 | NP_766100.1 | 9 | 1620 | |||
| GenBank mRNA | AK030849.1 | - | 9 | 1389 | ||
| AK036692.1 | - | 9 | 2201 | |||
| AK137604.1 | - | 2 | 2986 | |||
| AK141382.1 | - | 10 | 1691 | |||
| AK163329.1 | - | 3 | 556 | |||
| BC150160.1 | - | 9 | 1282 | 40141577 | ||
| BC150482.1 | - | 9 | 1282 | 40141578 | ||

Gabpb2
Genomic Map