siRNA Details
Gene Symbol: Fbxo42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172518.3 | NP_766106.2 | 10 | 5493 | ||
GenBank mRNA | BC058667.1 | - | 10 | 5483 | 6410012 | |
BQ177305.1 | - | 1 | 484 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fbxo42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172518.3 | NP_766106.2 | 10 | 5493 | ||
GenBank mRNA | BC058667.1 | - | 10 | 5483 | 6410012 | |
BQ177305.1 | - | 1 | 484 |