siRNA Details
| Gene Symbol: Nipal4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172524.3 | NP_766112.1 | 6 | 1042 | ||
| GenBank mRNA | AK035561.1 | - | 6 | 1043 | ||
| AK035736.1 | - | 6 | 1019 | |||
| BC139819.1 | - | 6 | 1033 | 40141293 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nipal4| Gene Symbol: Nipal4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172524.3 | NP_766112.1 | 6 | 1042 | ||
| GenBank mRNA | AK035561.1 | - | 6 | 1043 | ||
| AK035736.1 | - | 6 | 1019 | |||
| BC139819.1 | - | 6 | 1033 | 40141293 | ||