siRNA Details
| Gene Symbol: Lrrc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146048.1 | NP_001139520.1 | 7 | 861 | ||
| NM_172528.3 | NP_766116.3 | 7 | 509 | |||
| GenBank mRNA | AK051043.1 | - | 7 | 860 | ||
| AK082721.1 | - | 7 | 510 | |||
| AK147075.1 | - | 7 | 861 | |||
| AK220166.1 | - | 7 | 879 | |||
| BC046591.1 | - | 7 | 848 | |||
| BC087542.1 | - | 7 | 708 | 4934980 | ||

Lrrc1
Genomic Map