siRNA Details
| Gene Symbol: Aldh5a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172532.3 | NP_766120.1 | 3 | 527 | ||
| GenBank mRNA | AK052703.1 | - | 3 | 528 | ||
| AK144030.1 | - | 3 | 511 | |||
| AK159485.1 | - | 3 | 508 | |||
| AK166313.1 | - | 3 | 508 | |||
| BC138703.1 | - | 3 | 490 | 8861725 | ||

Aldh5a1
Genomic Map