siRNA Details
| Gene Symbol: Aldh5a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172532.3 | NP_766120.1 | 7 | 1124 | ||
| GenBank mRNA | AK052703.1 | - | 7 | 1125 | ||
| AK144030.1 | - | 7 | 1108 | |||
| AK159485.1 | - | 7 | 1105 | |||
| AK166313.1 | - | 7 | 1104 | |||
| BC138703.1 | - | 7 | 1087 | 8861725 | ||

Aldh5a1
Genomic Map