siRNA Details
| Gene Symbol: Rufy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172557.2 | NP_766145.1 | 5 | 771 | ||
| GenBank mRNA | AJ250024.1 | - | 4 | 447 | ||
| AK046625.1 | - | 5 | 788 | |||
| AK046633.1 | - | 5 | 772 | |||
| AK051178.1 | - | 5 | 777 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rufy1| Gene Symbol: Rufy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172557.2 | NP_766145.1 | 5 | 771 | ||
| GenBank mRNA | AJ250024.1 | - | 4 | 447 | ||
| AK046625.1 | - | 5 | 788 | |||
| AK046633.1 | - | 5 | 772 | |||
| AK051178.1 | - | 5 | 777 | |||