siRNA Details
| Gene Symbol: Rhbdf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001167680.1 | NP_001161152.1 | 11, 12 | 1616 | ||
| NM_172572.3 | NP_766160.2 | 11, 12 | 1830 | |||
| GenBank mRNA | AK028872.1 | - | 11, 12 | 1831 | ||
| AK154639.1 | - | 11, 12 | 1596 | |||
| AK171085.1 | - | 11, 12 | 1612 | |||
| AY167043.1 | - | 11, 12 | 1616 | |||
| BC031398.1 | - | 5, 6 | 649 | |||
| BC052182.1 | - | 11, 12 | 1564 | 6335022 | ||

Rhbdf2
Genomic Map