siRNA Details
| Gene Symbol: Engase | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172573.2 | NP_766161.1 | 14 | 3326 | ||
| GenBank mRNA | AK048667.1 | - | 14 | 3328 | ||
| BC060640.1 | - | 12 | 3696 | |||
| BC120923.1 | - | 14 | 3324 | |||
| BC120924.1 | - | 14 | 3328 | 40097903 | ||

Engase
Genomic Map