siRNA Details
| Gene Symbol: Tmem63c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172583.2 | NP_766171.1 | 11, 12 | 1024 | ||
| GenBank mRNA | AK030062.1 | - | 11, 12 | 1016 | ||
| AK034410.1 | - | 10, 11 | 947 | |||
| AK043536.1 | - | 10, 11 | 945 | |||
| BC056936.1 | - | 11, 12 | 1024 | 5695989 | ||
| BC057088.1 | - | 11, 12 | 1024 | 5695989 | ||

Tmem63c
Genomic Map