siRNA Details
| Gene Symbol: Wdr41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK082348.1 | - | 10 | 2071 | ||
| BC138095.1 | - | 10 | 1560 | 8861114 | ||
| BC145143.1 | - | 10 | 1610 | |||
| BC145144.1 | - | 10 | 1560 | 9053695 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wdr41| Gene Symbol: Wdr41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK082348.1 | - | 10 | 2071 | ||
| BC138095.1 | - | 10 | 1560 | 8861114 | ||
| BC145143.1 | - | 10 | 1610 | |||
| BC145144.1 | - | 10 | 1560 | 9053695 | ||