siRNA Details
| Gene Symbol: Wdhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172598.3 | NP_766186.2 | 9, 10 | 1203 | ||
| GenBank mRNA | BC063740.1 | - | 9, 10 | 1136 | 6328626 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wdhd1| Gene Symbol: Wdhd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172598.3 | NP_766186.2 | 9, 10 | 1203 | ||
| GenBank mRNA | BC063740.1 | - | 9, 10 | 1136 | 6328626 | |