siRNA Details
| Gene Symbol: Naprt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172607.3 | NP_766195.2 | 6 | 1058 | ||
| GenBank mRNA | AK033661.1 | - | 6 | 1061 | ||
| AK049157.1 | - | 6 | 807 | |||
| AK159127.1 | - | 6 | 857 | |||
| AK170518.1 | - | 6 | 857 | |||
| AY214331.1 | - | 6 | 1061 | |||
| BC058800.1 | - | 6 | 828 | 6476979 | ||

Naprt
Genomic Map