siRNA Details
| Gene Symbol: Gramd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205353.1 | NP_001192282.1 | 17 | 1831 | ||
| NM_172611.4 | NP_766199.1 | 17 | 1733 | |||
| GenBank mRNA | AK036832.1 | - | 17 | 1722 | ||
| AK129209.1 | - | 13 | 3708 | |||
| AK142266.1 | - | 16 | 2482 | |||
| BC021523.1 | - | 13 | 1107 | |||
| BC085148.1 | - | 17 | 1789 | |||
| BC139228.1 | - | 17 | 1722 | 8862250 | ||
| BC139229.1 | - | 17 | 1722 | 8862251 | ||

Gramd4
Genomic Map