siRNA Details
| Gene Symbol: Piezo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039485.4 | NP_001034574.4 | 46 | 8131 | ||
| GenBank mRNA | AK031235.1 | - | 6 | 1069 | ||
| AK031422.1 | - | 5 | 790 | |||
| BC026614.1 | - | 3 | 519 | |||
| BC075677.1 | - | 7 | 1222 | |||
| BC147606.1 | - | 5 | 684 | 9056596 | ||
| BC172109.1 | - | 5 | 684 | |||
| HQ215521.1 | - | 46 | 7211 | |||

Piezo2
Genomic Map