siRNA Details
| Gene Symbol: Piezo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK031235.1 | - | 8 | 3104 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Piezo2| Gene Symbol: Piezo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK031235.1 | - | 8 | 3104 | ||