siRNA Details
| Gene Symbol: Hectd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163471.1 | NP_001156943.1 | 21 | 2581 | ||
| NM_172637.3 | NP_766225.2 | 21 | 2578 | |||
| GenBank mRNA | AK029549.1 | - | 21 | 2438 | ||
| AK035511.1 | - | 7 | 808 | |||
| AK142777.1 | - | 19 | 2405 | |||
| BC137671.1 | - | 21 | 2552 | 8860690 | ||
| BC144815.1 | - | 21 | 2552 | 9053363 | ||

Hectd2
Genomic Map