siRNA Details
| Gene Symbol: Tmem194b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142647.1 | NP_001136119.1 | 8, 9 | 1177 | ||
| GenBank mRNA | AK030361.1 | - | 7, 8 | 1032 | ||
| AK036693.1 | - | 8, 9 | 1091 | |||
| AK048484.1 | - | 8, 9 | 1088 | |||
| AK086801.1 | - | 4, 5 | 574 | |||
| AK136177.1 | - | 4, 5 | 665 | |||
| AK170577.1 | - | 3, 4 | 2481 | |||
| BC028796.1 | - | 5, 6 | 603 | |||
| BC094370.1 | - | 3, 4 | 2121 | |||

Tmem194b
Genomic Map